missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrophenylhydrazine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 11452260
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100032-79-9
Molecular Formula C6H7BrClN3O2
Molecular Weight (g/mol) 268.50
MDL Number MFCD09260482
InChI Key SIBURLORIWZQNT-UHFFFAOYSA-N
Synonym 4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
PubChem CID 44119379
IUPAC Name (4-bromo-2-nitrophenyl)hydrazine;hydrochloride
SMILES Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O

Specifications

Melting Point ∽185°C (decomposition)
Quantity 1 g
Formula Weight 268.5
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-nitrophenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'