missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Borono-L-phenylalanine, 97%

Product Code. 10074103
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76410-58-7
Molecular Formula C9H12BNO4
Molecular Weight (g/mol) 209.01
MDL Number MFCD01075172
InChI Key NFIVJOSXJDORSP-QMMMGPOBSA-N
Synonym 4-borono-l-phenylalanine, s-2-amino-3-4-boronophenyl propanoic acid, 4-boronophenylalanine, p-boronophenylalanine, para-boronophenylalanine, 10b-bpa, unii-jsm63lv5md, para-borono-l-phenylalanine, l-phe b oh 2, l-4-boronophenylalanine
PubChem CID 150315
IUPAC Name (2S)-2-amino-3-(4-boronophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)B(O)O)C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 283.0°C to 293.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 209.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Borono-L-phenylalanine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Borono-L-phenylalanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'