missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15488517
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174132-31-1
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.567
MDL Number MFCD00237007
InChI Key KVUAOWDVYMUKPE-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxycarbonyl amino phenyl propanoic acid, fmoc-l-4-aminophe boc, fmoc-phe 4-nhboc-oh, fmoc-l-phe 4-nhboc-oh, 2s-3-4-tert-butoxycarbonyl amino phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-p-amino-phe boc-oh, ambotzfaa1741, fmoc-4-boc-amino-l-phenylalanine, s-fmoc-4-boc-amino phenylalanine, n-9h-fluorene-9-ylmethoxycarbonyl-4-tert-butoxycarbonylamino-l-phenylalanine
PubChem CID 2756117
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 5 g
Formula Weight 502.57
Percent Purity 95%
Chemical Name or Material 4-(Boc-amino)-N-Fmoc-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'