missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxycarbonylamino)benzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15429437
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192804-36-7
Molecular Formula C14H14BNO4
Molecular Weight (g/mol) 271.079
MDL Number MFCD02093055
InChI Key KYCQBMOJDXVNIC-UHFFFAOYSA-N
Synonym 4-benzyloxy carbonyl amino phenyl boronic acid, 4-benzyloxycarbonylamino phenylboronic acid, 4-cbz-aminophenyboronic acid, 4-cbz-aminophenyl boronic acid, 4-benzyloxy carbonyl amino phenylboronic acid, 4-benzyloxycarbonylamino-benzeneboronic acid, 4-aminobenzeneboronic acid, n-cbz protected, 4-benzyloxycarbonylamino-benzene boronic acid
PubChem CID 2773322
IUPAC Name [4-(phenylmethoxycarbonylamino)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)NC(=O)OCC2=CC=CC=C2)(O)O

Specifications

Melting Point 168°C to 169°C
Quantity 5 g
Formula Weight 271.08
Percent Purity 96%
Chemical Name or Material 4-(Benzyloxycarbonylamino)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'