missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-Fmoc-L-phenylalanine, 98%, Thermo Scientific Chemicals

Product Code. 15489028
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95753-56-3
Molecular Formula C24H22N2O4
Molecular Weight (g/mol) 402.45
MDL Number MFCD00237664
InChI Key VALNSJHHRPSUDO-QFIPXVFZSA-N
Synonym fmoc-4-amino-l-phenylalanine, fmoc-phe 4-nh2-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-aminophenyl propanoic acid, n, a-fmoc-4-amino-l-phenylalanine, fmoc-4-amino-phenylalanine, l-phenylalanine, 4-amino-n-9h-fluoren-9-ylmethoxy carbonyl, 2s-3-4-aminophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-4-ao-l-phenylalanine, fmoc-phe p-nh2-oh, pubchem12464
PubChem CID 7019927
IUPAC Name (2S)-3-(4-aminophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)N)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 402.44
Percent Purity 98%
Chemical Name or Material 4-Amino-N-Fmoc-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'