missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-bromopyrimidine-5-carbonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15429517
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94741-70-5
Molecular Formula C5H3BrN4
Molecular Weight (g/mol) 199.011
MDL Number MFCD00173661
InChI Key CXYLLFGNJJCGHM-UHFFFAOYSA-N
Synonym 4-amino-2-bromo-5-cyanopyrimidine, 4-amino-2-bromo-5-pyrimidinecarbonitrile, 5-pyrimidinecarbonitrile, 4-amino-2-bromo, 4-azanyl-2-bromanyl-pyrimidine-5-carbonitrile, 2-bromo-4-amino-5-cyanopyrimidine, acmc-209xix, 4-amino-2-bromo-pyrimidine-5-carbonitrile, 4-amino-2-bromopyrimidine-5-carbonitrile
PubChem CID 2735285
IUPAC Name 4-amino-2-bromopyrimidine-5-carbonitrile
SMILES C1=C(C(=NC(=N1)Br)N)C#N

Specifications

Melting Point 254°C (decomposition)
Quantity 250 mg
Formula Weight 199.01
Percent Purity 97%
Chemical Name or Material 4-Amino-2-bromopyrimidine-5-carbonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'