missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1H-pyrazole, 95%, Thermo Scientific Chemicals

Product Code. 11482180
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28466-26-4
Molecular Formula C3H5N3
Molecular Weight (g/mol) 83.094
MDL Number MFCD01693729
InChI Key AXINVSXSGNSVLV-UHFFFAOYSA-N
Synonym 4-aminopyrazole, 4-amino-1h-pyrazole, 1h-pyrazol-4-ylamine, 9ci, pyrazole, 4-amino, 1h-pyrazole-4-amine, pyrazole-4-ylamine, 4-nh2-pyrazole, 1-h-pyrazol-4-amine, pubchem14988
PubChem CID 78035
IUPAC Name 1H-pyrazol-4-amine
SMILES C1=C(C=NN1)N

Specifications

Melting Point 72°C to 77°C
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 83.09
Percent Purity 95%
Chemical Name or Material 4-Amino-1H-pyrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.