missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxybenzonitrile, 97%, Thermo Scientific™

Product Code. 15410678
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Acetoxybenzonitrile is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13031-41-9
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00129158
InChI Key CJGXWABHYYJNJH-UHFFFAOYSA-N
Synonym p-cyanophenyl acetate, benzonitrile, 4-acetyloxy, 4-acetoxybenzonitrile, unii-eel1pcp8am, eel1pcp8am, 4-cyanophenyl acetate, 4-acetyloxy benzonitrile, benzonitrile, p-hydroxy-, acetate ester
PubChem CID 83062
IUPAC Name (4-cyanophenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C#N

Specifications

Quantity 1 g
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 4-Acetoxybenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'