missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,5-Dimethoxy-o-phenylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2'-dithiobis(1-aminonaphthalene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).
Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.
Notes
Incompatible with oxidizing agents.
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).
Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | ∽265°C (decomposition) |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 3714004 |
Solubility Information | Soluble in warm methanol,dimethyl sulfoxide and water. |
IUPAC Name | 4,5-dimethoxybenzene-1,2-diamine;dihydrochloride |
Formula Weight | 241.12 |
Percent Purity | 98% |
Chemical Name or Material | 4,5-Dimethoxy-o-phenylenediamine dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'