missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenedianiline, SPEX CertiPrep™

Product Code. 16359327 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
4,4'-Methylenedianiline101-77-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 101-77-9, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C13H14N2
MDL Number MFCD00007914
InChI Key YBRVSVVVWCFQMG-UHFFFAOYSA-N
SMILES NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
Molecular Weight (g/mol) 198.27
PubChem CID 7577
ChEBI CHEBI:32506
Solvent or Matrix Methanol-P&T
Chemical Name or Material 4,4'-Methylenedianiline
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenedianiline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol, 4,4'-diaminodiphenylmethane

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.