missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)cyclohexanone, 97+%

Product Code. 15482108
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15482108

Brand: Thermo Scientific Alfa Aesar H55767.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105640-07-1 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105640-07-1
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00210693
InChI Key SLJYPZJZQIHNGU-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl cyclohexanone, 4-4-hydroxyphenyl cyclohexan-1-one, 4-4-hydroxyphenyl-cyclohexanone, cyclohexanone, 4-4-hydroxyphenyl, 4-4-hydroxyphenyl-1-cyclohexanone, acmc-1cukv, 4-4-oxocyclohexyl phenol, ksc495c5l, 4-4'-hydroxyphenyl-cyclohexanone
PubChem CID 4598916
IUPAC Name 4-(4-hydroxyphenyl)cyclohexan-1-one
SMILES OC1=CC=C(C=C1)C1CCC(=O)CC1

Specifications

Melting Point 170°C to 175°C
Quantity 5 g
Beilstein 3269332
Formula Weight 190.24
Percent Purity ≥97%
Chemical Name or Material 4-(4-Hydroxyphenyl)cyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.