missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diaminoazobenzene, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4,4'-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.
Solubility
Very soluble in ethanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
4,4′-Diaminoazobenzene is used in cross-linking studies of epoxy resins and the synthesis of surface relief holographic materials. It is used in synthesis of anthraquinone reactive dyes.
Solubility
Very soluble in ethanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | ∽245°C (decomposition) |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 745553 |
Merck Index | 14,2977 |
Solubility Information | Very soluble in ethanol. Insoluble in water. |
IUPAC Name | 4-[(4-aminophenyl)diazenyl]aniline |
Formula Weight | 212.26 |
Percent Purity | 95% |
Chemical Name or Material | 4,4'-Diaminoazobenzene |
Safety and Handling
Product Identifier
- 4,4'-Diaminoazobenzene
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'