Learn More
4,4'-Bipyridine, 98%
Description
4,4'-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N'-dimethyl-4,4'-bipyridinium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,4′-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N′-dimethyl-4,4′-bipyridinium.
Solubility
Soluble in ethanol, ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | 111°C to 114°C |
| Boiling Point | 304°C to 305°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 113176 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,3348 |
| Solubility Information | Soluble in ethanol,ether. Insoluble in water. |
| Formula Weight | 156.19 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
- 4,4'-Bipyridine
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- MIXTURE LIST-Contains : 4,4-Bipyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.