missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Trifluoromethyl)phenoxy]piperidine, 99%, Thermo Scientific™

Product Code. 11434810
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824390-04-7
Molecular Formula C12H14F3NO
Molecular Weight (g/mol) 245.245
MDL Number MFCD06248881
InChI Key MBZJEIRMMYDAFD-UHFFFAOYSA-N
Synonym 4-2-trifluoromethyl phenoxy piperidine, piperidine, 4-2-trifluoromethyl phenoxy, 4-2-trifluoromethyl-phenoxy-piperidine, 4-2-trifluoromethylphenoxy piperidine, acmc-20aiak, 4-2-trifluoromethyl-phqsy piperidine, 4-2-trifluoromethyl phenoxy-piperidine
PubChem CID 5018790
IUPAC Name 4-[2-(trifluoromethyl)phenoxy]piperidine
SMILES C1CNCCC1OC2=CC=CC=C2C(F)(F)F

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 245.25
Percent Purity 99%
Chemical Name or Material 4-[2-(Trifluoromethyl)phenoxy]piperidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.