missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15448945
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112539-09-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.704
MDL Number MFCD00219329
InChI Key OUAVTZYGHGCCCB-UHFFFAOYSA-N
PubChem CID 2794593
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 104°C to 105°C
Quantity 5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Synonym 4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl
Formula Weight 259.71
Percent Purity 97%
Chemical Name or Material 4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'