Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
528.00€ - 554.00€
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Material | Polystyrene |
No. of Wells | 384 |
Sterility | Non-sterile |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11866091
|
Greiner Bio-One
781900 |
Black |
528.00€
Pack of 40 |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11893891
|
Greiner Bio-One
781904 |
White |
554.00€
Pack of 40 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
Description
- Free of detectable DNase, RNase, human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Higher assay sensitivity with reduced background
Service Plan
No
Specifications
Standard | |
Polystyrene | |
Non-sterile |
16 x 24 | |
384 |