Learn More
Applied Biosystems™ Plate Septa, 384 well
These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.
Brand: Applied Biosystems™ 4315934
Includes: Each package contains 20 septa, sufficient for 7,680 samples
Description
Specifications
Each package contains 20 septa, sufficient for 7,680 samples. | |
3100-Avant Genetic Analyzer | |
20 each |
384-well Plate | |
Septa Plate | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.