Learn More
Applied Biosystems™ Plate Septa, 384 well
These are septa for use with the 3730, 3730xl, 3130xl, 3130, 3100 and 3100-Avant™ instruments.
Brand:  Applied Biosystems™ 4315934
Product Code. 16385272
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Includes: Each package contains 20 septa, sufficient for 7,680 samples
Specifications
Each package contains 20 septa, sufficient for 7,680 samples.
3100-Avant Genetic Analyzer
20 each
384-well Plate
Septa Plate
Room Temperature