Learn More
Fisherbrand™ Stainless Steel Chattaway Spatulas
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
100 mm
300 mm
350 mm
Unit Size:
Pack of 5
Description
Scrapes and lifts materials from bottles or flasks. Nickel Electro™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish. Can be used for sterile applications.
- Manufactured from 304 18/8 Stainless Steel
- Rounded bent end is useful for scraping or lifting materials from bottles or flasks
- Non-magnetic
- Bright finish
Specifications
Specifications
| Quantity | 5 Pk. |
| Sterility | Sterile |
| Autoclavable | Autoclavable |
| Product Type | Chattaway Spatula |
| Finish | Bright |
| For Use With (Application) | For Sterile Applications |
| Width (Metric) Blade | 10 mm |
| Temperature (Metric) Max | -196°C to +550°C |
| Material | Stainless Steel |
| Length (Metric) | 300 mm |
Product Content Correction
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur