missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzoyl chloride, ≥97%, Thermo Scientific™

Product Code. 10263193
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10263193

Brand: Thermo Scientific Maybridge CC57102DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3586-15-0
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
MDL Number MFCD03424712
InChI Key TTZXIWBOKOZOPL-UHFFFAOYSA-N
Synonym benzoyl chloride, 3-phenoxy, m-phenoxybenzoyl chloride, m-phenoxy benzoylchoride, 3-phenoxy benzoyl chloride, 3-phenoxy-benzoyl chloride, benzoyl chloride,3-phenoxy
PubChem CID 2760341
IUPAC Name 3-phenoxybenzoyl chloride
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 232.67
Percent Purity 97%
Chemical Name or Material 3-Phenoxybenzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.