missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 11382239
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 796061-08-0
Molecular Formula C19H24BNO4S
Molecular Weight (g/mol) 373.274
MDL Number MFCD05663863
InChI Key RGAZOXRZFCGICF-UHFFFAOYSA-N
Synonym 4-methyl-n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 3-p-toluenesulfonylamino phenylboronic acid pinacol ester, 3-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 3-toluene-4-sulfonylamino phenylboronic acid pinacol ester, n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-tolueneulfonamide, 3-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 3-toluene-4-sulfonylamino phenyl boronic acid, pinacol ester
PubChem CID 16217721
IUPAC Name 4-methyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NS(=O)(=O)C3=CC=C(C=C3)C

Specifications

Melting Point 164°C to 168°C
Quantity 1 g
Formula Weight 373.27
Percent Purity 97%
Chemical Name or Material 3-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'