missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrothiobenzamide, 97%, Thermo Scientific Chemicals

Product Code. 15478677
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70102-34-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD09757586
InChI Key HDQCHDWHHGEXQE-UHFFFAOYSA-N
Synonym 3-nitrobenzene-1-carbothioamide, 3-nitrothiobenzamide, 3-nitrobenzothioamide, 3-nitro-thiobenzamide, 3-nitrobenzenethioamide, 3-nitro-thio-benzamide, benzenecarbothioamide, 3-nitro
PubChem CID 12668182
IUPAC Name 3-nitrobenzenecarbothioamide
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=S)N

Specifications

Quantity 1 g
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 3-Nitrothiobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'