missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 15473887
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86256-63-5
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.15
MDL Number MFCD09025380
InChI Key NCXWNLUZPMBTHX-UHFFFAOYSA-N
Synonym 3-methyl-5-trifluoromethoxy aniline, 5-methyl-3-trifluoromethoxy phenylamine, 3-methyl-5-trifluoromethoxy aniline, jrd
PubChem CID 46737546
IUPAC Name 3-methyl-5-(trifluoromethoxy)aniline
SMILES CC1=CC(N)=CC(OC(F)(F)F)=C1

Specifications

Boiling Point 73°C (1 mmHg)
Refractive Index 1.4692
Quantity 250 mg
UN Number UN2810
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethoxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'