missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorostyrene, 97%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Product Code. 11419583
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350-51-6
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00000341
InChI Key ZJSKEGAHBAHFON-UHFFFAOYSA-N
Synonym 3-fluorostyrene, 1-fluoro-3-vinylbenzene, m-fluorostyrene, benzene, 1-ethenyl-3-fluoro, 1-ethenyl-3-fluoro-benzene, 3-fluoro-styrene, acmc-1crku, 1-fluoro-3-vinyl-benzene, 1-fluoro-3-vinylbenzene #, 3-fluorostyrene, contains 4-tert-butylcatechol as inhibitor
PubChem CID 67691
IUPAC Name 1-ethenyl-3-fluorobenzene
SMILES C=CC1=CC(=CC=C1)F

Specifications

Boiling Point 40°C to 42°C (8 mmHg)
Flash Point 29°C (84°F)
Odor Recognizable
Quantity 1 g
UN Number UN1993
Beilstein 2038488
Formula Weight 122.14
Percent Purity 97%
Chemical Name or Material 3-Fluorostyrene, Stabilized with 0.1% 4-tert-butylcatechol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'