missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-D-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15419658
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110117-84-5
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00066449
InChI Key VWHRYODZTDMVSS-UHFFFAOYNA-N
Synonym 3-fluoro-d-phenylalanine, d-3-fluorophenylalanine, h-d-phe 3-f-oh, 2r-2-amino-3-3-fluorophenyl propanoic acid, m-fluoro-d-phenylalanine, 3-fluoro-d-phe, d-3-fluorophe, r-2-amino-3-3-fluorophenyl propanoic acid, 3-fluor-d-phenylalanin, h-m-fluoro-d-phe-oh
PubChem CID 716317
SMILES NC(CC1=CC(F)=CC=C1)C(O)=O

Specifications

IUPAC Name (2R)-2-amino-3-(3-fluorophenyl)propanoic acid
Formula Weight 183.18
Percent Purity 95%
Quantity 25 g
Physical Form Solid
Chemical Name or Material 3-Fluoro-D-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'