missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylphenol, 95%, Thermo Scientific Chemicals

Product Code. 11456420
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-17-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002311
InChI Key HMNKTRSOROOSPP-UHFFFAOYSA-N
Synonym m-ethylphenol, phenol, 3-ethyl, phenol, m-ethyl, meta-ethylphenol, 1-ethyl-3-hydroxybenzene, 1-hydroxy-3-ethylbenzene, 3-ethyl-phenol, unii-0g9zk222jx, benzene, 1-ethyl-3-hydroxy, dsstox_cid_2480
PubChem CID 12101
ChEBI CHEBI:34332
IUPAC Name 3-ethylphenol
SMILES CCC1=CC=CC(O)=C1

Specifications

Melting Point -4°C
Density 1.028
Boiling Point 218°C
Flash Point 94°C (202°F)
Refractive Index 1.5335
Quantity 250 g
UN Number UN3145
Beilstein 1857002
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 3-Ethylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'