missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15438507
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80126-51-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD01075124
InChI Key JJDJLFDGCUYZMN-QMMMGPOBSA-N
Synonym 3-chloro-l-phenylalanine, l-3-chlorophenylalanine, s-2-amino-3-3-chlorophenyl propanoic acid, h-phe 3-cl-oh, 2s-2-amino-3-3-chlorophenyl propanoic acid, 3-chlorophenylalanine, l-phe 3-cl-oh, 3-chloro-l-phenylalanine hydrochloride, d-3-chlorophenylalanine, l-3-chlorophe
PubChem CID 2761492
ChEBI CHEBI:49597
IUPAC Name (2S)-2-amino-3-(3-chlorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(Cl)=C1)C(O)=O

Specifications

Formula Weight 199.64
Percent Purity 95%
Quantity 250 mg
Chemical Name or Material 3-Chloro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'