missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzylamine, 97%, Thermo Scientific™

Product Code. 11376779
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90390-33-3
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.59
MDL Number MFCD06213787
InChI Key XJCYOVBALKWQQC-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzylamine, 3-chloro-5-fluorophenyl methanamine, benzenemethanamine, 3-chloro-5-fluoro, 3-chloro-5-fluoro benzylamine, 1-3-chloro-5-fluorophenyl methanamine, 3-chloro-5-fluoro-benzylamine, 3-chloro-5-fluorobenzyl amine, 5-chloro-3-fluorophenyl methylamine, 3-chloro-5-fluoro-phenyl methanamine, 3-chloro-5-fluoro-benzenemethanamine
PubChem CID 457616
SMILES NCC1=CC(F)=CC(Cl)=C1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
IUPAC Name (3-chloro-5-fluorophenyl)methanamine
Formula Weight 159.59
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'