missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11409676
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65055-17-6
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00236215
InChI Key DLZUHSFUBZTBQZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
PubChem CID 2736548
IUPAC Name 3-chloro-4-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)F

Specifications

Boiling Point 222°C
Refractive Index 1.551
Quantity 5 g
UN Number UN3265
Beilstein 6288904
Sensitivity Moisture sensitive
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'