Learn More
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%, Thermo Scientific Chemicals
76.80€ - 323.00€
Chemical Identifiers
CAS | 78266-06-5 |
---|---|
Molecular Formula | C15H19BrN2O5 |
Molecular Weight (g/mol) | 387.23 |
MDL Number | MFCD00216974 |
InChI Key | MHPZZZZLAQGTHT-UHFFFAOYSA-N |
Synonym | mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine |
PubChem CID | 54158 |
IUPAC Name | 2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid |
SMILES | CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382877
|
Thermo Scientific Alfa Aesar
L16086.03 |
1 g |
76.80€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392877
|
Thermo Scientific Alfa Aesar
L16086.06 |
5 g |
323.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
78266-06-5 | |
387.23 | |
MHPZZZZLAQGTHT-UHFFFAOYSA-N | |
54158 | |
CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C |
C15H19BrN2O5 | |
MFCD00216974 | |
mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine | |
2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid |
Specifications
78266-06-5 | |
C15H19BrN2O5 | |
mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine | |
MHPZZZZLAQGTHT-UHFFFAOYSA-N | |
2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid | |
54158 | |
≥98% |
192°C to 196°C | |
MFCD00216974 | |
Soluble in dimethylsulphoxide and ethanol. Insoluble in water. | |
CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C | |
387.23 | |
387.24 | |
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 278-877-6
missing translation for 'rtecsNumber' : MB9121850
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only