missing translation for 'onlineSavingsMsg'
Learn More

3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals

Product Code. 11307826
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1455-42-1
Molecular Formula C15H28O6
Molecular Weight (g/mol) 304.383
MDL Number MFCD00059794
InChI Key BPZIYBJCZRUDEG-UHFFFAOYSA-N
Synonym 2,2'-2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diyl bis 2-methylpropan-1-ol, 3,9-bis 1,1-dimethyl-2-hydroxyethyl-2,4,8,10-tetraoxaspiro 5.5 undecane, cyclic acetal, 2-9-1-hydroxy-2-methylpropan-2-yl-2,4,8,10-tetraoxaspiro 5.5 undecan-3-yl-2-methylpropan-1-ol, acmc-209cve, ksc489m7h, 2,4,8,10-tetraoxaspiro 5.5 undecane-3,9-diethanol, beta3,beta3,beta9,beta9-tetramethyl, bis hydroxypivalaldehyde pentaerythritolacetal, bis hydroxypivalaldehyde pentaerythritolacetal cyclic acetal, bis hydroxypivalaldehyde pentaerythritol acetal cyclic acetal
PubChem CID 7569008
IUPAC Name 2-[3-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]-2-methylpropan-1-ol
SMILES CC(C)(CO)C1OCC2(CO1)COC(OC2)C(C)(C)CO

Specifications

Melting Point 197°C to 200°C
Quantity 25 g
Beilstein 1574570
Formula Weight 304.39
Percent Purity 97%
Chemical Name or Material 3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'