missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Difluoroacetophenone, 98%, Thermo Scientific™

Product Code. p-3759038 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10786475

Brand: Thermo Scientific Acros 398120010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 123577-99-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00042489
InChI Key OXJLDNSPGPBDCP-UHFFFAOYSA-N
Synonym 3',5'-difluoroacetophenone, 3,5-difluoroacetophenone, 1-3,5-difluorophenyl ethanone, 1-3,5-difluorophenyl ethan-1-one, ethanone, 1-3,5-difluorophenyl, 1-acetyl-3,5-difluorobenzene, 1-3,5-difluoro-phenyl-ethanone, pubchem2179, acmc-1cacd, 3,5-di-fluoroacetophenone
PubChem CID 518596
IUPAC Name 1-(3,5-difluorophenyl)ethanone
SMILES CC(=O)C1=CC(F)=CC(F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3COCH3
Quantity 1 g
Formula Weight 156.13
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 3', 5'-Difluoroacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3',5'-Difluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.