missing translation for 'onlineSavingsMsg'
Learn More

3,4-Methylenedioxy-beta-nitrostyrene, 98%, Thermo Scientific™

Product Code. 11397246
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1485-00-3
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00014575
InChI Key KFLWBZPSJQPRDD-ONEGZZNKSA-N
Synonym 3,4-methylenedioxy-beta-nitrostyrene, mns, syk inhibitor iii, 1,3-benzodioxole, 5-nitrovinyl, 5-2-nitrovinyl benzo d 1,3 dioxole, 1,3-benzodioxole, 5-2-nitroethenyl, 5-2-nitrovinyl-1,3-benzodioxole, 5-2-nitroethenyl-1,3-benzodioxole, styrene, 3,4-methylenedioxy-beta-nitro, e-5-2-nitrovinyl benzo d 1,3 dioxole
PubChem CID 672296
IUPAC Name 5-[(E)-2-nitroethenyl]-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)C=C[N+](=O)[O-]

Specifications

Melting Point 159°C to 163°C
Quantity 10 g
Beilstein 192350
Sensitivity Light sensitive
Formula Weight 193.16
Percent Purity 98%
Chemical Name or Material 3,4-Methylenedioxy-beta-nitrostyrene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'