Learn More
3-(4-Fluorophenyl)-1-isopropylindole, 98%, Thermo Scientific™
Product Code. 11329115
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93957-49-4
Molecular Formula C17H16FN
Molecular Weight (g/mol) 253.32
MDL Number MFCD01075733
InChI Key ZDZJOIIBECYKAJ-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
PubChem CID 736229
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole
SMILES CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F
Specifications
Melting Point 95°C to 98°C
Quantity 1 g
Beilstein 4457093
Formula Weight 253.32
Percent Purity 98%
Chemical Name or Material 3-(4-Fluorophenyl)-1-isopropylindole