Learn More
3-(2-Carboxyethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Product Code. 15429747
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 693803-17-7
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.993
MDL Number MFCD04115649
InChI Key JJBQHOJBARDWRZ-UHFFFAOYSA-N
Synonym 3-3-boronophenyl propanoic acid, 3-2-carboxyethyl phenylboronic acid, 3-3-boronophenyl propionic acid, 3-2-carboxyethyl phenyl boronic acid, benzenepropanoic acid, 3-borono-9ci, 3-3-dihydroxyboranyl phenyl propanoic acid, 3-2-carboxyethyl benzeneboronic acid, acmc-1bfcs, 3-carboxyethylphenylboronic acid
PubChem CID 3856118
IUPAC Name 3-(3-boronophenyl)propanoic acid
SMILES B(C1=CC(=CC=C1)CCC(=O)O)(O)O
Specifications
Melting Point 166°C to 169°C
Quantity 250 mg
Formula Weight 193.99
Percent Purity 96%
Chemical Name or Material 3-(2-Carboxyethyl)benzeneboronic acid