missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15419285
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257651-17-4
Molecular Formula C19H29BN2O5
Molecular Weight (g/mol) 376.26
MDL Number MFCD16294500
InChI Key IYTPRTWBNLJZLK-UHFFFAOYSA-N
Synonym 3-2-boc-amino acetamido benzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl-carbamic acid tert-butyl ester, tert-butyl n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamoyl methyl carbamate, amtb154, 3-2-boc-amino acetamido phenylboronic acid pinacol ester, 3-2-tert-butoxycarbonylamino acetamido phenylboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylcarbamoyl-methyl carbamic acid tert-butyl ester, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenylcarbamoyl methyl carbamic acid tert-butyl ester, tert-butyl 2-oxo-2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anilino ethyl carbamate
PubChem CID 57415693
IUPAC Name tert-butyl N-[2-oxo-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anilino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCC(=O)NC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Formula Weight 376.26
Percent Purity 97%
Chemical Name or Material 3-[2-(Boc-amino)acetamido]benzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'