Learn More
3-(2,4-Dichlorophenyl)propanoic acid, 97%, Thermo Scientific™
9.80€ - 33.60€
Chemical Identifiers
CAS | 55144-92-8 |
---|---|
Molecular Formula | C9H8Cl2O2 |
Molecular Weight (g/mol) | 219.061 |
MDL Number | MFCD00275605 |
InChI Key | HLNPVFSCAMKIOD-UHFFFAOYSA-N |
PubChem CID | 2740915 |
IUPAC Name | 3-(2,4-dichlorophenyl)propanoic acid |
SMILES | C1=CC(=C(C=C1Cl)Cl)CCC(=O)O |
Description
Chemical Identifiers
55144-92-8 | |
219.061 | |
HLNPVFSCAMKIOD-UHFFFAOYSA-N | |
3-(2,4-dichlorophenyl)propanoic acid |
C9H8Cl2O2 | |
MFCD00275605 | |
2740915 | |
C1=CC(=C(C=C1Cl)Cl)CCC(=O)O |
Specifications
55144-92-8 | |
97% | |
C9H8Cl2O2 | |
MFCD00275605 | |
HLNPVFSCAMKIOD-UHFFFAOYSA-N | |
3-(2,4-dichlorophenyl)propanoic acid | |
2740915 | |
97% |
97.0 | |
Amber Glass Bottle | |
Cl2C6H3CH2CH2CO2H | |
3-2,4-dichlorophenyl propanoic acid, 3-2,4-dichlorophenyl propionic acid, 2,4-dichlorohydrocinnamic acid, 2,4-dichloro-benzenepropanoic acid, benzenepropanoic acid, 2,4-dichloro, 3-2,4-dichloro-phenyl-propionic acid, maybridge4_004135, acmc-1b21h, ksc603c0n, benzenepropanoic acid,2,4-dichloro | |
C1=CC(=C(C=C1Cl)Cl)CCC(=O)O | |
219.061 | |
219.07 | |
3-(2, 4-Dichlorophenyl)propanoic acid |
Sicherheit und Handhabung
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.