missing translation for 'onlineSavingsMsg'
Learn More

Mepacrine Hydrochloride, Mikromol™

Product Code. 30051587
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30051587

Brand: Mikromol MM1695.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Mepacrine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mepacrine Hydrochloride
CAS 69-05-6
Analyte or Component Names Mepacrine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C23 H30 Cl N3 O . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Mepacrine Hydrochloride, 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, hydrochloride (1:2), 1,4-Pentanediamine, N4-(6-chloro-2-methoxy-9-acridinyl)-N1,N1-diethyl-, dihydrochloride (9CI), Acridine, 6-chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxy-, dihydrochloride (8CI), 2-Chloro-5-(ω-diethylamino-α-methylbutylamino)-7-methoxyacridine dihydrochloride, 2-Methoxy-6-chloro-9-(4-diethylamino-1-methylbutylamino)acridine dihydrochloride, 3-Chloro-7-methoxy-9-(1-methyl-4-diethylaminobutylamino)acridine dihydrochloride, 3-Chloro-9-(4'-diethylamino-1'-methylbutylamino)-7-methoxyacridine dihydrochloride, 6-Chloro-9-[[4-(diethylamino)-1-methylbutyl]amino]-2-methoxyacridine dihydrochloride, 866RP, Acrichin dihydrochloride, Atabrine hydrochloride, Atebrin, Atebrine, Atebrine hydrochloride, Chemiochin, Chinacrin hydrochloride, Crinodora, Erion, Malaricida, Mecryl, Mepacrine dihydrochloride, Mepacrine hydrochloride, Methoquine, Metochin, Metoquin, Metoquine, NSC 14229, Palacrin, Palusan, Pentilen, Quinacrine dihydrochloride, Quinacrine hydrochloride, SN 390
InChI Formula InChI=1S/C23H30ClN3O.2ClH/c1-5-27(6-2)13-7-8-16(3)25-23-19-11-9-17(24)14-22(19)26-21-12-10-18(28-4)15-20(21)23;;/h9-12,14-16H,5-8,13H2,1-4H3,(H,25,26);2*1H
SMILES Cl.Cl.CCN(CC)CCCC(C)Nc1c2ccc(Cl)cc2nc3ccc(OC)cc13
IUPAC Name 4-N-(6-chloro-2-methoxyacridin-9-yl)-1-N,1-N-diethylpentane-1,4-diamine;dihydrochloride
Molecular Weight (g/mol) 472.88
Quantity 250 mg
Formula Weight 471.1611 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.