Learn More
Fisherbrand™ Nickel Microspatula
Use with blances, microscopes and weighing boats. The Fisherbrand™ Nickel Microspatula is designed to dispense and transfer samples during weighing.
Brand:  Fisherbrand™ FB65055
Product Code. 11716447
  • 69.65€ / Pack of 5
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dispense and transfer samples during weighing
  • Apply micro amounts of adhesive for tacking down specimens, spreading paste and mixing granular samples
  • Flat-ended blade
Specifications
Specifications
5 Pk.
Bright
4 mm
Nickel
Spatula
For Structural Applications Where Resistance to Corrosion is Prime Consideration
315°C
130 mm