Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand: RETSCH 60.123.000710
This item is not returnable.
View return policy
Specifications
1Each | |
Test Sieve | |
Stainless Steel | |
25 mm | |
200 mm | |
Stainless Steel |
710 μm | |
ASTM E 11 | |
Shaker AS 300 | |
For separation, fractioning, particle size determination | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve > Pore Size: 710 μm