Learn More
2-(Trimethylsilyl)thiophene, 97%
Product Code. p-7041162
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15437678
Brand: Thermo Scientific Alfa Aesar H61170.06
Request Bulk or Custom Format
CAS: 18245-28-8 | C7H12SSi | 156.318 g/mol
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.
Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18245-28-8
Molecular Formula C7H12SSi
Molecular Weight (g/mol) 156.318
MDL Number MFCD00005415
InChI Key OANGLGSZPSFVDY-UHFFFAOYSA-N
Synonym 2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
PubChem CID 140360
IUPAC Name trimethyl(thiophen-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CS1
Specifications
Density 0.926
Flash Point 41°C (106°F)
Refractive Index 1.497
Quantity 5 g
UN Number UN1993
Formula Weight 156.32
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiophene