missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)pyridine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.
Solubility
Soluble in most organic solvents.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Lithiation with n-BuLi/LiOCH2CH2NMe2 takes place at the 6-position, affording access to 6-substituted derivatives. With LiTMP . It is a building block in heterocycle synthesis.
Solubility
Soluble in most organic solvents.
Notes
Store at room temperature. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.28 |
Boiling Point | 139°C to 141°C |
Flash Point | 42°C (108°F) |
Refractive Index | 1.418 |
Quantity | 250 mg |
UN Number | UN1993 |
Beilstein | 3112 |
Solubility Information | Soluble in most organic solvents. |
Formula Weight | 147.1 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'