missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)phenylacetic acid, 98%, Thermo Scientific™

Product Code. 11460378
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220239-67-8
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD00236325
InChI Key QFLBBLZROQPTAI-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenylacetic acid, 2-2-trifluoromethoxy phenyl acetic acid, 2-trifluoromethoxy phenyl acetic acid, benzeneacetic acid, 2-trifluoromethoxy, pubchem7379, acmc-209wg4, ksc495e8p, rarechem al bo 0857, 2-trifluoromethoxyphenylacetic acid
PubChem CID 2777313
IUPAC Name 2-[2-(trifluoromethoxy)phenyl]acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)OC(F)(F)F

Specifications

Melting Point 54°C to 56°C
Quantity 1 g
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethoxy)phenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'