missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP, Honeywell Riedel-de Haën™

Product Code. 15676890
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.784 - 0.786 g/cm3
Boiling Point 77°C to 83°C
Quantity 1 L
Flash Point 12°C
Packaging Plastic Bottle
UN Number UN1219
Beilstein 635639
Formula Weight 60.11
Percent Purity >99.9%
Grade ACS Reagent
Chemical Name or Material 2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'