missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propoxybenzaldehyde, 97+%, Thermo Scientific™

Product Code. 11365529
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7091-12-5
PubChem CID 249807

Specifications

Quantity 1 g
Molecular Formula C4H8O
MDL Number MFCD00005356
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
Formula Weight 164.2
Percent Purity ≥97%
Chemical Name or Material 2-n-Propoxybenzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'