missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propoxybenzaldehyde, 97+%, Thermo Scientific™

Product Code. 11355529
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7091-12-5
PubChem CID 249807

Specifications

Molecular Formula C10H12O2
MDL Number MFCD00046150
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 2-propoxy-benzaldehyde, 2-propyloxybenzaldehyde, 2-n-propoxybenzaldehyde, propoxy benzaldehyde, acmc-20anok, pubchem3160, benzaldehyde,-2-propoxy, benzaldehyde,2-propoxy, aronis23744, 2,6-dichloro-4-nitropyridine 3-oxide
InChI Key CDUPASLURGOXGD-UHFFFAOYSA-N
SMILES CCCOC1=CC=CC=C1C=O
IUPAC Name 2-propoxybenzaldehyde
Molecular Weight (g/mol) 164.20
Formula Weight 164.2
Percent Purity ≥97%
Chemical Name or Material 2-n-Propoxybenzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'