missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylthioadenosine diphosphate trisodium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Potent purinergic agonist displaying selectivity for P2Y1, P2Y12 and P2Y13 receptors (pEC50 = 8.29 and 9.05 for P2Y1 and P2Y12, EC50 = 19 nM for P2Y13). Induces aggregation of, and inhibits cAMP accumulation in, platelets in vitro.

Specifications
Specifications
CAS | 475193-31-8 |
Molecular Formula | C11H17N5Na3O10P2S |
Quantity | 10 mg |
Synonym | 2-methylthioadenosine diphosphate trisodium salt |
InChI Key | BIJRFSXCZAGVHQ-MSQVLRTGSA-N |
SMILES | CSC1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na] |
IUPAC Name | [(2R,3S,4R,5R)-5-(6-amino-2-methylsulfanylpurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium |
Molecular Weight (g/mol) | 542.259 |
PubChem CID | 131854177 |
Formula Weight | 539.24 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction