missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dithiane, 98%, Thermo Scientific Chemicals

Product Code. 11362705
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6007-26-7
Molecular Formula C5H10S2
Molecular Weight (g/mol) 134.255
MDL Number MFCD00014652
InChI Key KXROTPXCYDXGSC-UHFFFAOYSA-N
Synonym 1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane
PubChem CID 22329
IUPAC Name 2-methyl-1,3-dithiane
SMILES CC1SCCCS1

Specifications

Density 1.212
Boiling Point 78°C to 80°C (9 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.5615
Quantity 1 g
Beilstein 102766
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 134.25
Percent Purity 98%
Chemical Name or Material 2-Methyl-1,3-dithiane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'