missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, Honeywell

Product Code. 15603980
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Density 0.966-0.968g/mL at 20°C, 0.965g/mL at 25°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Linear Formula CH3OCH2CH2OH
Quantity 1 L
UN Number UN1188
Beilstein 1731074
Formula Weight 76.09g/mol
Vapor Pressure 6.17 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'