missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4-phenylthiazole, 98%, Thermo Scientific Chemicals

Product Code. 11347605
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-88-0
Molecular Formula C9H7NS2
Molecular Weight (g/mol) 193.282
MDL Number MFCD00014508
InChI Key CYCKHTAVNBPQDB-UHFFFAOYSA-N
Synonym 4-phenylthiazole-2-thiol, 2-mercapto-4-phenylthiazole, 4-phenyl-1,3-thiazole-2-thiol, 4-phenyl-2-thiazolethiol, 2 3h-thiazolethione, 4-phenyl, 4-phenyl-2-mercaptothiazole, 4-thiazoline-2-thione, 4-phenyl, 2-thiazolethiol, 4-phenyl, 4-phenyl-thiazole-2-thiol, 4-phenylthiazole-2 3h-thione
PubChem CID 3000729
IUPAC Name 4-phenyl-3H-1,3-thiazole-2-thione
SMILES C1=CC=C(C=C1)C2=CSC(=S)N2

Specifications

Melting Point 172°C to 175°C
Odor Odorless
Quantity 5 g
Beilstein 128288
Formula Weight 193.3
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4-phenylthiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'