missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycarbazole, 97%, Thermo Scientific Chemicals

Product Code. 11382929
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-79-3
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00004962
InChI Key GWPGDZPXOZATKL-UHFFFAOYSA-N
Synonym 2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole
PubChem CID 93551
IUPAC Name 9H-carbazol-2-ol
SMILES OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Specifications

Melting Point 262°C to 273°C
Odor Odorless
Quantity 25 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxycarbazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.